Merck
  • Involvement of an antioxidant defense system in the adaptive response to cadmium in maize seedlings (Zea mays L.).
Involvement of an antioxidant defense system in the adaptive response to cadmium in maize seedlings (Zea mays L.).
Bulletin of environmental contamination and toxicology (2014-08-27)
Xianghua Xu, Cuiying Liu, Xiaoyan Zhao, Renying Li, Wenjing Deng
ABSTRACT
Chemical and biological analyses were used to investigate the growth response and antioxidant defense mechanism of maize seedlings (Zea mays L.) grown in soils with 0-100 mg kg(-1) Cd. Results showed that maize seedlings have strong abilities to accumulate and tolerate high concentrations of Cd. For soil with 50 mg kg(-1) Cd, the Cd contents in roots and shoots of maize seedlings are as large as 295.6 and 153.0 mg kg(-1) DW, respectively, without visible symptoms of toxicity. Lower soil Cd concentrations lead to a decrease in reduced glutathione (GSH) content in leaves of maize seedlings, whereas higher soil Cd concentrations resulted in an increase in the activities of superoxide dismutase, guaiacol peroxidase, catalase, and ascorbate peroxidase. Maize seedlings have strong capacities to adapt to low concentrations of Cd by consuming GSH and to develop an antioxidative enzyme system to defend against high-Cd stress.
Product Description
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, foil, 200x200mm, thickness 1.0mm, as rolled, 99.99%
Cadmium, foil, 10mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 30x50mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, wire reel, 5m, diameter 0.125mm, as drawn, 99.9%
Cadmium, foil, 15mm disks, thickness 0.015mm, 99.7+%
Cadmium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9%
Cadmium, wire reel, 5m, diameter 0.5mm, hard, 99.99+%
Cadmium, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 15mm disks, thickness 0.01mm, 99.7+%
Cadmium, wire reel, 2m, diameter 0.5mm, hard, 99.95+%
Cadmium, foil, 4mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 25mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 15mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, 50x50mm, thickness 1.0mm, as rolled, 99.95%
Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%
Cadmium, foil, 1m coil, thickness 0.5mm, as rolled, 100%
Cadmium, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%
Cadmium, foil, 10mm disks, thickness 0.25mm, as rolled, 99.95%