Merck
  • History of bioavailable lead and iron in the Greater North Sea and Iceland during the last millennium - a bivalve sclerochronological reconstruction.
History of bioavailable lead and iron in the Greater North Sea and Iceland during the last millennium - a bivalve sclerochronological reconstruction.
Marine pollution bulletin (2014-09-02)
Hilmar A Holland, Bernd R Schöne, Soraya Marali, Klaus P Jochum
ABSTRACT
We present the first annually resolved record of biologically available Pb and Fe in the Greater North Sea and Iceland during 1040-2004 AD based on shells of the long-lived marine bivalve Arctica islandica. The iron content in pre-industrial shells from the North Sea largely remained below the detection limit. Only since 1830, shell Fe levels rose gradually reflecting the combined effect of increased terrestrial runoff of iron-bearing sediments and eutrophication. Although the lead gasoline peak of the 20th century was well recorded by the shells, bivalves that lived during the medieval heyday of metallurgy showed four-fold higher shell Pb levels than modern specimens. Presumably, pre-industrial bivalves were offered larger proportions of resuspended (Pb-enriched) organics, whereas modern specimens receive fresh increased amounts of (Pb-depleted) phytoplankton. As expected, metal loads in the shells from Iceland were much lower. Our study confirms that bivalve shells provide a powerful tool for retrospective environmental biomonitoring.
Product Description
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Lead, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 500x500mm, thickness 0.5mm, as rolled, 99.95%
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Lead, foil, 150x150mm, thickness 0.15mm, as rolled, 99.95%
Iron, rod, 25mm, diameter 10.0mm, as drawn, 99.99+%
Iron, rod, 100mm, diameter 100mm, as drawn, armcO« soft ingot 99.8%
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, rod, 100mm, diameter 30mm, as drawn, armcO« soft ingot 99.8%
Iron, rod, 100mm, diameter 2.0mm, as drawn, 99.95%
Iron, foil, 25x25mm, thickness 1.0mm, as rolled, 99.5%
Iron, foil, 100x100mm, thickness 2.0mm, as rolled, 99.95%
Iron, foil, 300x300mm, thickness 0.1mm, hard, 99.5%
Iron, wire reel, 1m, diameter 0.5mm, as drawn, 99.95%
Iron, foil, 5m coil, thickness 0.1mm, hard, 99.5%
Iron, foil, 10m coil, thickness 0.25mm, hard, 99.5%
Iron, rod, 50mm, diameter 5.0mm, as drawn, 99.99+%
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Lead, foil, 2m coil, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.12mm, coil width 25.4mm, as rolled, 99.95%
Iron, wire reel, 5m, diameter 0.5mm, hard, 99.99+%
Iron, rod, 100mm, diameter 30mm, as drawn, 98+%
Iron, rod, 500mm, diameter 19mm, as drawn, soft ingot 99.8%
Iron, rod, 200mm, diameter 50mm, as drawn, 98+%
Iron, rod, 300mm, diameter 6.0mm, as drawn, 99.99+%
Iron, tube, 1000mm, outside diameter 12.0mm, inside diameter 9mm, wall thickness 1.5mm, annealed, 99.5%
Iron, rod, 100mm, diameter 40mm, as drawn, armcO« soft ingot 99.8%