Merck
  • Concentrations of lead, cadmium and barium in urban garden-grown vegetables: the impact of soil variables.
Concentrations of lead, cadmium and barium in urban garden-grown vegetables: the impact of soil variables.
Environmental pollution (Barking, Essex : 1987) (2014-08-29)
Murray B McBride, Hannah A Shayler, Henry M Spliethoff, Rebecca G Mitchell, Lydia G Marquez-Bravo, Gretchen S Ferenz, Jonathan M Russell-Anelli, Linda Casey, Sharon Bachman
ABSTRACT
Paired vegetable/soil samples from New York City and Buffalo, NY, gardens were analyzed for lead (Pb), cadmium (Cd) and barium (Ba). Vegetable aluminum (Al) was measured to assess soil adherence. Soil and vegetable metal concentrations did not correlate; vegetable concentrations varied by crop type. Pb was below health-based guidance values (EU standards) in virtually all fruits. 47% of root crops and 9% of leafy greens exceeded guidance values; over half the vegetables exceeded the 95th percentile of market-basket concentrations for Pb. Vegetable Pb correlated with Al; soil particle adherence/incorporation was more important than Pb uptake via roots. Cd was similar to market-basket concentrations and below guidance values in nearly all samples. Vegetable Ba was much higher than Pb or Cd, although soil Ba was lower than soil Pb. The poor relationship between vegetable and soil metal concentrations is attributable to particulate contamination of vegetables and soil characteristics that influence phytoavailability.
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Barium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Barium, rod, diam. ~2 cm, ≥99% trace metals basis
Sigma-Aldrich
Barium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Barium, beads, 0.5-2.0 mm particle size, 99% trace metals basis
Sigma-Aldrich
Barium, pieces, 1 cm, 99% trace metals basis
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, rod, 500mm, diameter 5.0mm, as drawn, 99.99+%
Lead, foil, 2m coil, thickness 0.016mm, as rolled, 96% (sn 1.5%, sb 2.5%)
Cadmium, foil, not light tested, 25x25mm, thickness 0.001mm, 99.7+%
Lead, foil, 4mm disks, thickness 0.009mm, 99.99+%
Cadmium, foil, 8mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, tube, 200mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Lead, foil, 25mm disks, thickness 1.0mm, as rolled, 99.99+%
Lead, foil, 0.5m coil, thickness 1.0mm, as rolled, 99.95%
Lead, foil, 50mm disks, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 1.0mm, as rolled, 99.99+%
Lead, foil, 50x50mm, thickness 0.5mm, as rolled, 99.99+%
Lead, foil, 10mm disks, thickness 1.0mm, as rolled, 99.99+%
Lead, foil, 1m coil, thickness 0.05mm, as rolled, 97% (sn 1.5%, sb 1.5%)
Lead, foil, 300x300mm, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 10m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 0.5m coil, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.01mm, 99.99+%
Lead, foil, 0.5m coil, thickness 1.0mm, as rolled, 99.5%
Lead, foil, 8mm disks, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 6mm disks, thickness 0.5mm, as rolled, 99.95%