Merck
  • Cellular genomic reporter assays for screening and evaluation of inducers of fetal hemoglobin.
Cellular genomic reporter assays for screening and evaluation of inducers of fetal hemoglobin.
Human molecular genetics (2003-12-03)
Jim Vadolas, Hady Wardan, Michael Orford, Robert Williamson, Panayiotis A Ioannou
ABSTRACT
Reactivation of fetal hemoglobin (HbF) expression using pharmacological agents represents a potential strategy for the therapy of beta-thalassemia, sickle cell disease, HbE and other beta-hemoglobinopathies. However, the drugs currently available have low efficacy and specificity and are associated with high toxicity. We describe the development of stable cellular genomic reporter assays (GRAs) based on the green fluorescence protein (EGFP) gene under the Ggamma-globin promoter in the intact human beta-globin locus. We show that human erythroleukemic cell lines stably transfected with a Ggamma-EGFP beta-globin locus construct can maintain a uniform basal level of EGFP expression over long periods of continuous culture and that induction of EGFP expression parallels the induction of the endogenous globin genes. We compared the EGFP-induction potency of a number of chemotherapeutic agents, including histone deacetylase inhibitors and DNA-binding agents. We show that hydroxyurea and butyrate result in moderate levels of induction (70-80%) but with an additive inductive effect. Among the DNA-binding agents tested, cisplatin was the most potent inducer of HbF expression, (442+/-32%), a level which is comparable to hemin (764+/-145%). These results indicate that cellular GRAs containing Ggamma-EGFP-modified beta-globin locus constructs can be used to develop novel inducers of HbF synthesis for the therapy of beta-hemoglobinopathies.
Sigma-Aldrich
α-Methylhydrocinnamic acid, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Pricing and availability is not currently available.