Merck
  • Hydrogenolysis of 2,2-dimethylbutane and n-hexane over supported ruthenium, nickel, cobalt, and iron.
Hydrogenolysis of 2,2-dimethylbutane and <I>n</I>-hexane over supported ruthenium, nickel, cobalt, and iron.
J. Catal. (1979)
Machiels CJ and Anderson RB.