Merck
  • Calf uterine estradiol receptor. Effects of molybdate on salt-induced transformation process and characterization of a nontransformed receptor state.
Calf uterine estradiol receptor. Effects of molybdate on salt-induced transformation process and characterization of a nontransformed receptor state.
The Journal of biological chemistry (1981-11-25)
G Redeuilh, C Secco, E E Baulieu, H Richard-Foy
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein