Merck
  • Quantification of neurotoxin BMAA (β-N-methylamino-L-alanine) in seafood from Swedish markets.
Quantification of neurotoxin BMAA (β-N-methylamino-L-alanine) in seafood from Swedish markets.
Scientific reports (2014-11-07)
Liying Jiang, Nadezda Kiselova, Johan Rosén, Leopold L Ilag
ABSTRACT
The neurotoxin β-N-methylamino-L-alanine (BMAA) produced naturally by cyanobacteria, diatoms and dinoflagellates can be transferred and accumulated up the food chain, and may be a risk factor for neurodegenerative diseases. This study provides the first systematic screening of BMAA exposure of a large population through the consumption of seafood sold in metropolitan markets. BMAA was distinguished from known isomers by liquid chromatography tandem mass spectrometry after acidic hydrolysis and derivatization. Using deuterium-labeled internal standard, BMAA was quantified as 0.01-0.90 μg/g wet weight of tissues in blue mussel, oyster, shrimp, plaice, char and herring, but was undetectable (<0.01 μg/g) in other samples (salmon, cod, perch and crayfish). Provided that the content of BMAA detected is relevant for intake calculations, the data presented may be used for a first estimation of BMAA exposure through seafood from Swedish markets, and to refine the design of future toxicological experiments and assessments.
Sigma-Aldrich
L-2,4-Diaminobutyric acid dihydrochloride, ≥95.0%
Pricing and availability is not currently available.