Merck
  • Poly [[tris (di-2-pyridylamine) tris (μ-5-sulfonatosalicylato) tricopper (II)] trihydrate].
Poly [[tris (di-2-pyridylamine) tris (μ-5-sulfonatosalicylato) tricopper (II)] trihydrate].
Acta Crystallographica Section E, Structure Reports Online (2004)
Fan SR and Zhu LG.