Merck
  • Preliminary communication Properties of nonlinear supramolecular liquid crystals containing thiophenedicarboxylic acids.
Preliminary communication Properties of nonlinear supramolecular liquid crystals containing thiophenedicarboxylic acids.
Liq. Cryst. (1998)
Lin H-C, et al.