Merck
  • Development of thyroglobulin antibodies after GVAX immunotherapy is associated with prolonged survival.
Development of thyroglobulin antibodies after GVAX immunotherapy is associated with prolonged survival.
International journal of cancer (2014-05-17)
Alessandra De Remigis, Tanja D de Gruijl, Jennifer N Uram, Schey-Cherng Tzou, Shintaro Iwama, Monica V Talor, Todd D Armstrong, Saskia J A M Santegoets, Susan F Slovin, Lei Zheng, Daniel A Laheru, Elizabeth M Jaffee, Winald R Gerritsen, Alfons J M van den Eertwegh, Dung T Le, Patrizio Caturegli
ABSTRACT
Cancer immunotherapy induces a variety of autoinflammatory responses, including those against the thyroid gland, which can be exploited to predict clinical outcomes. Considering the paucity of information about thyroid autoimmunity in patients receiving cancer vaccines, we designed our study to assess the development of thyroglobulin antibodies (TgAbs) in patients treated with GVAX (vaccine made of a tumor cell type transfected with GM-CSF) and/or ipilimumab and correlated seroconversion with survival. Using both in house and commercial ELISA assays, we measured TgAbs in patients with pancreatic (No. = 53), prostate (No. = 35) or colon (No. = 8) cancer, before and after treatment with GVAX only (No. = 34), GVAX plus ipilimumab (No. = 42) or ipilimumab (No. = 20), and correlated their levels with patient's survival, disease status and T-cell surface markers. Antibodies to thyroperoxidase, myeloperoxidase, proteinase 3, insulin and actin were also measured. TgAbs specifically developed after GVAX, independent of the underlying cancer (81% in prostate, 75% colon cancer and 76% pancreatic cancer) and co-administration of ipilimumab (75% in GVAX only and 78% in GVAX plus ipilimumab). This TgAbs seroconversion could be detected mainly by the in house assay, suggesting that the thyroglobulin epitopes recognized by the antibodies induced by GVAX are different from the epitopes seen in the classic form of Hashimoto thyroiditis. Notably, TgAbs seroconversion was associated with significantly prolonged survival (p = 0.01 for pancreas and p = 0.005 for prostate cancer). In conclusion, GVAX immunotherapy induces the appearance of TgAbs that recognize a unique antigenic repertoire and associate with prolonged survival.
Sigma-Aldrich
Thyroglobulin from bovine thyroid, powder, ≥90% (agarose gel electrophoresis)
Sigma-Aldrich
Thyroglobulin from bovine thyroid, (For Gel Filtration Chromatography)
Sigma-Aldrich
Thyroglobulin human
Sigma-Aldrich
Thyrotropic hormone from bovine pituitary
Sigma-Aldrich
Thyroglobulin from porcine thyroid gland, powder
Sigma-Aldrich
Thyrotropic hormone from human pituitary, lyophilized powder, ≥6.2 IU/mg (immunoassay, non-sterile)