Merck
  • A comparison of in vitro ADME properties and pharmacokinetics of azithromycin and selected 15-membered ring macrolides in rodents.
A comparison of in vitro ADME properties and pharmacokinetics of azithromycin and selected 15-membered ring macrolides in rodents.
European journal of drug metabolism and pharmacokinetics (2013-10-12)
Astrid Milić, Vlatka Bencetić Mihaljević, Jovica Ralić, Ana Bokulić, Danijela Nožinić, Branka Tavčar, Boris Mildner, Vesna Munić, Ivica Malnar, Jasna Padovan
ABSTRACT
The purpose of this study was to evaluate the impact of structural modifications on the 15-membered macrolactone ring and/or substituents on the in vitro ADME properties and in vivo pharmacokinetic (PK) profile for selected derivatives in rodents in comparison to azithromycin. Azithromycin and seven selected 15-membered macrolide derivatives, modified either by removal of the sugar moieties, replacement of the amine with a lactam, or addition of lipophilic substituents, were screened in several in vitro ADME assays and in vivo PK studies in rodents. In vitro ADME profiling included assessment of passive permeability and P-gp substrate, metabolic stability in liver microsomes and hepatocytes, as well as CYP direct inhibition measurements. In vivo PK studies were performed in rats (Sprague-Dawley), mice (Balb/c), and P-gp wild-type and deficient mice (CF-1™). Different structural modifications on the azithromycin scaffold resulted in substantial changes in disposition kinetics and oral bioavailability in both rodent species. However, these differences in vivo cannot be predicted based on in vitro results since most of these molecules are classified in the same category. Therefore, in the case of 15-membered ring macrolides, the in vitro ADME screens presented here seem to have low predictive value for in vivo prediction, making their use as routine in vitro screens prior to PK assessments questionable.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Azithromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Azithromycin for system suitability, European Pharmacopoeia (EP) Reference Standard
Azithromycin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
7-Ethoxycoumarin
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture