Merck
  • First line and salvage therapy with total therapy 3-based treatment for multiple myeloma- an extended single center experience.
First line and salvage therapy with total therapy 3-based treatment for multiple myeloma- an extended single center experience.
Leukemia research (2014-07-26)
Eli Muchtar, Ron Ram, Pia Raanani, Moshe Yeshurun, Nino Oniashvili, Ofer Shpilberg, Hila Magen
ABSTRACT
Total therapy 3 is an intensified protocol for multiple myeloma (MM). The "real life" outcomes of this protocol were seldom reported. Data was obtained for 81 patients (newly diagnosed, n=49; progressive MM, n=32), most of which had high-risk parameters. Overall response rate following (V)DT-PACE was 96% and 75% for the newly diagnosed and progressive groups, respectively. Median progression-free survival was 42.5 and 9 months, respectively. The 2-year overall survival was 88% and 40%, respectively. Treatment with (V)DT-PACE achieves high response rate among patients with high-risk disease, which can be translated into long-term remission only for newly diagnosed patients.
Product Description
Etoposide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(±)-Thalidomide, ≥98%, powder
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard