Merck
  • Isotopic ratio based source apportionment of children's blood lead around coking plant area.
Isotopic ratio based source apportionment of children's blood lead around coking plant area.
Environment international (2014-08-16)
Suzhen Cao, Xiaoli Duan, Xiuge Zhao, Beibei Wang, Jin Ma, Delong Fan, Chengye Sun, Bin He, Fusheng Wei, Guibin Jiang
ABSTRACT
Lead exposure in the environment is a major hazard affecting human health, particularly for children. The blood lead levels in the local children living around the largest coking area in China were measured, and the source of blood lead and the main pathways of lead exposure were investigated based on lead isotopic ratios ((207)Pb/(206)Pb and (208)Pb/(206)Pb) in blood and in a variety of media, including food, airborne particulate matter, soil, dust and drinking water. The children's blood lead level was 5.25 (1.59 to 34.36 as range) μg dL(-1), lower than the threshold in the current criteria of China defined by the US Centers for Disease Control (10 μg dL(-1)). The isotopic ratios in the blood were 2.111±0.018 for (208)Pb/(206)Pb and 0.864±0.005 for (207)Pb/(206)Pb, similar to those of vegetables, wheat, drinking water, airborne particulate matter, but different from those of vehicle emission and soil/dust, suggesting that the formers were the main pathway of lead exposure among the children. The exposure pathway analysis based on the isotopic ratios and the human health risk assessment showed that dietary intake of food and drinking water contributed 93.67% of total exposed lead. The study further indicated that the coal used in the coking plant is the dominant pollution source of lead in children's blood.
Product Description
Lead, foil, light tested, 50x50mm, thickness 0.015mm, 99.99+%
Lead, foil, not light tested, 50x50mm, thickness 0.004mm, 99.99+%
Lead, foil, not light tested, 100x100mm, thickness 0.04mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, not light tested, 100x100mm, thickness 0.007mm, 99.99+%
Lead, foil, not light tested, 100x100mm, thickness 0.006mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.03mm, 99.99+%
Lead, rod, 100mm, diameter 6.35mm, 99.95%
Lead, foil, not light tested, 100x100mm, thickness 0.009mm, 99.99+%
Lead, foil, not light tested, 150x150mm, thickness 0.03mm, 99.99+%
Lead, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.015mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.025mm, 99.99+%
Lead, foil, not light tested, 50x50mm, thickness 0.015mm, 99.99+%
Lead, wire reel, 0.5m, diameter 0.75mm, as drawn, 99.99+%
Lead, rod, 1000mm, diameter 12.7mm, 99.95%
Lead, foil, not light tested, 150x150mm, thickness 0.05mm, 99.99+%
Lead, tube, 1000mm, outside diameter 6.78mm, inside diameter 4.7mm, wall thickness 1.04mm, 99.9%
Lead, microfoil, disks, 25mm, thinness 0.25μm, specific density 266.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Lead, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.99+%
Lead, rod, 200mm, diameter 50mm, 99.95%
Lead, microfoil, disks, 10mm, thinness 0.25μm, specific density 266.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Lead, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.99+%
Lead, wire reel, 1m, diameter 1.0mm, as drawn, 99.99+%
Lead, tube, 500mm, outside diameter 6.78mm, inside diameter 4.7mm, wall thickness 1.04mm, 99.9%
Lead, foil, light tested, 50x50mm, thickness 0.02mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.016mm, as rolled, 96% (Sn 1.5%, Sb 2.5%)
Lead, foil, light tested, 25x25mm, thickness 0.009mm, 99.99+%
Lead, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%