Merck
  • Treatment of hypertension and renal injury induced by the angiogenesis inhibitor sunitinib: preclinical study.
Treatment of hypertension and renal injury induced by the angiogenesis inhibitor sunitinib: preclinical study.
Hypertension (Dallas, Tex. : 1979) (2014-09-04)
Stephanie Lankhorst, Mariëtte H W Kappers, Joep H M van Esch, Frank M M Smedts, Stefan Sleijfer, Ron H J Mathijssen, Hans J Baelde, A H Jan Danser, Anton H van den Meiracker
ABSTRACT
Common adverse effects of angiogenesis inhibition are hypertension and renal injury. To determine the most optimal way to prevent these adverse effects and to explore their interdependency, the following drugs were investigated in unrestrained Wistar Kyoto rats exposed to the angiogenesis inhibitor sunitinib: the dual endothelin receptor antagonist macitentan; the calcium channel blocker amlodipine; the angiotensin-converting enzyme inhibitor captopril; and the phosphodiesterase type 5 inhibitor sildenafil. Mean arterial pressure was monitored telemetrically. After 8 days, rats were euthanized and blood samples and kidneys were collected. In addition, 24-hour urine samples were collected. After sunitinib start, mean arterial pressure increased rapidly by ≈30 mm Hg. Coadministration of macitentan or amlodipine largely prevented this rise, whereas captopril or sildenafil did not. Macitentan, captopril, and sildenafil diminished the sunitinib-induced proteinuria and endothelinuria and glomerular intraepithelial protein deposition, whereas amlodipine did not. Changes in proteinuria and endothelinuria were unrelated. We conclude that in our experimental model, dual endothelin receptor antagonism and calcium channel blockade are suitable to prevent angiogenesis inhibition-induced hypertension, whereas dual endothelin receptor antagonism, angiotensin-converting enzyme inhibitor, and phosphodiesterase type 5 inhibition can prevent angiogenesis inhibition-induced proteinuria. Moreover, the variable response of hypertension and renal injury to different antihypertensive agents suggests that these side effects are, at least in part, unrelated.
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder
Sigma-Aldrich
Amlodipine besylate, ≥98% (HPLC)
Amlodipine for peak identification, European Pharmacopoeia (EP) Reference Standard
Amlodipine besylate, European Pharmacopoeia (EP) Reference Standard
USP
Amlodipine besylate, United States Pharmacopeia (USP) Reference Standard
Supelco
Amlodipine besylate, Pharmaceutical Secondary Standard; Certified Reference Material