Merck
  • The making of a sweet modification: structure and function of O-GlcNAc transferase.
The making of a sweet modification: structure and function of O-GlcNAc transferase.
The Journal of biological chemistry (2014-10-23)
John Janetzko, Suzanne Walker
ABSTRACT
O-GlcNAc transferase is an essential mammalian enzyme responsible for transferring a single GlcNAc moiety from UDP-GlcNAc to specific serine/threonine residues of hundreds of nuclear and cytoplasmic proteins. This modification is dynamic and has been implicated in numerous signaling pathways. An unexpected second function for O-GlcNAc transferase as a protease involved in cleaving the epigenetic regulator HCF-1 has also been reported. Recent structural and biochemical studies that provide insight into the mechanism of glycosylation and HCF-1 cleavage will be described, with outstanding questions highlighted.
Sigma-Aldrich
Phosphorylase b from rabbit muscle, lyophilized powder, ≥20 units/mg protein, 2× crystallization
Sigma-Aldrich
Phosphorylase b from rabbit muscle, For use as a marker in SDS-PAGE