Merck
Short-term lung function decline in tunnel construction workers.
Occupational and environmental medicine (2014-11-02)
Bente Ulvestad, May Brit Lund, Berit Bakke, Yngvar Thomassen, Dag G Ellingsen
ABSTRACT
Tunnel construction workers are exposed to particulate and gaseous air contaminants. Previous studies carried out in the 1990s showed that tunnel construction workers were at increased risk of both short-term and long-term lung function decline. Since then, efforts have been made to reduce exposure. The objective of the present study was to investigate if current exposure may still cause short-term lung function impairment. Tunnel workers work 12 days consecutively, and then they are off for 9 days. Ninety tunnel workers and 51 referents were examined with spirometry and questionnaires before their work period started and again 11 days later. Personal exposure to particles and α-quartz in the thoracic aerosol subfraction, elemental carbon and organic carbon, oil mist, nitrogen dioxide and ammonia was assessed on two consecutive days between the two health examinations. The geometric means air concentrations for particulate matter in the thoracic mass aerosol subfraction, α-quartz, oil mist, organic carbon and elemental carbon for all workers were 561, 63, 210, 146 and 35 μg/m(3), respectively. After 11 days of work, the mean forced expiratory volume in 1 s (FEV1) in healthy participants had declined 73 mL (SD 173), p<0.001 in the tunnel workers, compared to 3 mL (SD 21), p=0.9 in the referents. Also, forced vital capacity (FVC) had declined significantly. Declines in FVC and FEV1 were significantly associated with exposure to organic carbon. In spite of reduced levels of exposure in modern tunnelling operations, a negative impact on lung function was still observed.
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Carbon, rod, 150mm, diameter 5.0mm, graphite, 100%
Carbon, foil, 12.5x12.5mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, rod, 100mm, diameter 3.0mm, graphite, 100%
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Millipore
Activated charcoal, suitable for GC
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size