Merck
  • Utilization of biochar sorbents for Cd²⁺, Zn²⁺, and Cu²⁺ ions separation from aqueous solutions: comparative study.
Utilization of biochar sorbents for Cd²⁺, Zn²⁺, and Cu²⁺ ions separation from aqueous solutions: comparative study.
Environmental monitoring and assessment (2014-11-20)
Vladimír Frišták, Martin Pipíška, Juraj Lesný, Gerhard Soja, Wolfgang Friesl-Hanl, Alena Packová
ABSTRACT
The objective of this study was to study the utilization of two different woody-derived biochars for Cd(2+), Zn(2+), and Cu(2+) ions separation from aqueous solutions. Physicochemical characterization confirmed the main differences in sorbent surface area and cation-exchange capacity. The maximum cadmium, zinc, and copper sorption capacities were 1.99, 0.97, and 2.50 mg g(-1) for biochar (BC) A; 7.80, 2.23, and 3.65 mg g(-1) for BC B. Sorption processes can be affected by time and pH. The most of sorbed cadmium and zinc were bound on exchangeable fractions and copper oxidizable fractions. Chemical modification and FT-IR analyses confirmed the crucial roles of hydroxyl and mainly carboxyl functional groups in sorption processes of Cd(2+), Zn(2+), and Cu(2+) ions by BC A and BC B. The garden wood rests with leaf mass-derived biochar can be utilized as an effective sorbent for bivalent ions.
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Copper, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Copper, IRMM®, certified reference material, 0.1 mm foil
Copper (S, P), BCR®, certified reference material, chips
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Copper, shot, −3-+14 mesh, 99%
Sigma-Aldrich
Copper, powder, 99.99% trace metals basis
Sigma-Aldrich
Zinc, purum, powder
Copper, IRMM®, certified reference material, 0.5 mm wire
Copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Cadmium, foil, not light tested, 25x25mm, thickness 0.01mm, 99.7+%
Zinc, foil, not light tested, 50x50mm, thickness 0.005mm, 99.9%
Zinc, wire reel, 50m, diameter 3.0mm, extruded, 99.9%
Copper - O.F.H.C., wire reel, 50m, diameter 0.125mm, annealed, 99.95+%
Copper - O.F.H.C., tube, 200mm, outside diameter 4.0mm, inside diameter 3mm, wall thickness 0.5mm, 99.95+%
Zinc, wire reel, 1m, diameter 0.5mm, as drawn, 99.99+%
Zinc, wire reel, 0.2m, diameter 1.0mm, 99.999%
Copper - O.F.H.C., tube, 1000mm, outside diameter 4.0mm, inside diameter 3mm, wall thickness 0.5mm, 99.95+%
Copper - O.F.H.C., wire reel, 10m, diameter 2.0mm, hard, 99.95+%