Merck
  • Endogenous S-sulfhydration of PTEN helps protect against modification by nitric oxide.
Endogenous S-sulfhydration of PTEN helps protect against modification by nitric oxide.
Biochemical and biophysical research communications (2014-12-03)
Kazuki Ohno, Kosaku Okuda, Takashi Uehara
ABSTRACT
Hydrogen sulfide (H2S) is a gaseous regulatory factor produced by several enzymes, and plays a pivotal role in processes such as proliferation or vasodilation. Recent reports demonstrated the physiological and pathophysiological functions of H2S in neurons. PTEN is a target of nitric oxide (NO) or hydrogen peroxide, and the oxidative modification of cysteine (Cys) residue(s) attenuates its enzymatic activity. In the present study, we assessed the effect of H2S on the direct modification of PTEN and the resulting downstream signaling. A modified biotin switch assay in SH-SY5Y human neuroblastoma cells revealed that PTEN is S-sulfhydrated endogenously. Subsequently, site-directed mutagenesis demonstrated that both Cys71 and Cys124 in PTEN are targets for S-sulfhydration. Further, the knockdown of cystathionine β-synthetase (CBS) using siRNA decreased this modification in a manner that was correlated to amount of H2S. PTEN was more sensitive to NO under these conditions. These results suggest that the endogenous S-sulfhydration of PTEN via CBS/H2S plays a role in preventing the S-nitrosylation that would inhibition its enzymatic activity under physiological conditions.
Sigma-Aldrich
sulfur powder, 99.998% trace metals basis
Supelco
Sulfur, PESTANAL®, analytical standard
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, 97%
SAFC
L-Cysteine
Sigma-Aldrich
Sulfur, powder, 99.98% trace metals basis
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Biotin, ≥99% (HPLC), lyophilized powder
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Biotin, European Pharmacopoeia (EP) Reference Standard
USP
Biotin, United States Pharmacopeia (USP) Reference Standard
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
BIOTIN