Merck
  • Bioavailability of cobalt and iron from citric-acid-adsorbed CoFe2O4 nanoparticles in the terrestrial isopod Porcellio scaber.
Bioavailability of cobalt and iron from citric-acid-adsorbed CoFe2O4 nanoparticles in the terrestrial isopod Porcellio scaber.
The Science of the total environment (2014-12-02)
Tea Romih, Barbara Drašler, Anita Jemec, Damjana Drobne, Sara Novak, Miha Golobič, Darko Makovec, Robert Susič, Ksenija Kogej
ABSTRACT
The aim of this study was to determine whether citric acid adsorbed onto cobalt ferrite (CoFe2O4) nanoparticles (NPs) influences the bioavailability of their constituents Co and Fe. Dissolution of Co and Fe was assessed by two measures: (i) in aqueous suspension using chemical analysis, prior to application onto the food of test organisms; and (ii) in vivo, measuring the bioavailability in the model terrestrial invertebrate (Porcellio scaber, Isopoda, Crustacea). The isopods were exposed to citric-acid-adsorbed CoFe2O4 NPs for 2 weeks, and tissue accumulation of Co and Fe was assessed. This was compared to pristine CoFe2O4 NPs, and CoCl2 and Fe(III) salts as positive controls. The combined data shows that citric acid enhances free metal ion concentration from CoFe2O4 NPs in aqueous suspension, although in vivo, very similar amounts of assimilated Co were found in isopods exposed to both types of NPs. Therefore, evaluation of the dissolution in suspension by chemical means is not a good predictor of metal assimilation of this model organism; body assimilation of Co and Fe is rather governed by the physiological capacity of P. scaber for the uptake of these metals. Moreover, we propose that citric acid, due to its chelating properties, may hinder the uptake of Co that dissolves from citric-acid-adsorbed CoFe2O4 NPs, if citric acid is present in sufficient quantity.
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Iron, IRMM®, certified reference material, 0.5 mm wire
Cobalt, foil, not light tested, 25x25mm, thickness 0.009mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.0125mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.05mm, annealed, 99.9%
Iron, foil, 15mm disks, thickness 0.38mm, hard, 99.5%
Cobalt, foil, not light tested, 100x100mm, thickness 0.006mm, 99.9%
Cobalt, foil, 25mm disks, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, 6mm disks, thickness 0.25mm, as rolled, 99.9%
Cobalt, foil, light tested, 50x50mm, thickness 0.0125mm, 99.9%
Cobalt, microfoil, disks, 10mm, thinness 0.25μm, specific density 222μg/cm2, permanent mylar 3.5μm support, 99.9%
Cobalt, foil, 4mm disks, thickness 0.008mm, 99.9%
Iron, foil, 10mm disks, thickness 0.075mm, hard, 99.5%
Cobalt, foil, not light tested, 25x25mm, thickness 0.01mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.009mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.015mm, 99.9%
Iron, foil, 25mm disks, thickness 0.003mm, 99.85%
Cobalt, foil, 8mm disks, thickness 0.05mm, as rolled, 99.9%