Merck
  • Vinculin negatively regulates transcription of MT1-MMP through MEK/ERK pathway.
Vinculin negatively regulates transcription of MT1-MMP through MEK/ERK pathway.
Biochemical and biophysical research communications (2014-12-03)
Taisuke Yoshimoto, Takahisa Takino, Zichen Li, Takahiro Domoto, Hiroshi Sato
ABSTRACT
Vinculin regulates a variety of cellular functions partly through stabilization of tumor suppressor PTEN. In order to study the role of vinculin in tumor progression other than PTEN stabilization, vinculin was knocked down in PTEN-deficient squamous cell carcinoma HSC-4 cells. Knockdown of vinculin induced phenotypical change by reducing cell-cell and cell-extracellular matrix adhesions, and enhanced MT1-MMP expression at transcription level and subsequent cell migration. Up-regulation of MT1-MMP transcription by vinculin knockdown was abrogated by ERK inhibition. These results suggest that vinculin negatively regulates malignant phenotype of tumor cells including MT1-MMP transcription through MEK/ERK pathway.
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Supelco
2-Butanone, analytical standard
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Supelco
Methyl Ethyl Ketone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder