Merck
  • Anti-smoking legislation and its effects on urinary cotinine and cadmium levels.
Anti-smoking legislation and its effects on urinary cotinine and cadmium levels.
Environmental research (2014-12-03)
Jinny E Sánchez-Rodríguez, Mónica Bartolomé, Ana I Cañas, Olga Huetos, Carmen Navarro, A Carolina Rodríguez, Misericordia Arribas, Marta Esteban, Ana López, Argelia Castaño
ABSTRACT
Anti-smoking legislation has been associated with an improvement in health indicators. Since the cadmium (Cd) body burden in the general population is markedly increased by smoke exposure, we analyzed the impact of the more restrictive legislation that came into force in Spain in 2011 by measuring Cd and cotinine in first morning urine samples from 83 adults in Madrid (Spain) before (2010) and after (2011) introduction of this law. Individual pair-wise comparisons showed a reduction of creatinine corrected Cotinine and Cd levels for non-active smokers, i. e. those which urinary cotinine levels are below 50 μg/L. After the application of the stricter law, cotinine levels in urine only decreased in non-active smokers who self-reported not to be exposed to second-hand smoke. The reduction in second hand smoke exposure was significantly higher in weekends (Friday to Sunday) than in working days (Monday to Thursday). The decrease in U-Cd was highly significant in non-active smokers and, in general, correlated with lower creatinine excretion. Therefore correction by creatinine could bias urinary Cd results, at least for cotinine levels higher than 500 μg/L. The biochemical/toxicological benefits detected herein support the stricter application of anti-smoking legislation and emphasize the need to raise the awareness of the population as regards exposure at home.
Sigma-Aldrich
(−)-Cotinine, ≥98%
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Supelco
(−)-Cotinine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Cadmium, foil, not light tested, 25x25mm, thickness 0.01mm, 99.7+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%
Cadmium, foil, 15mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, 50x50mm, thickness 1.0mm, as rolled, 99.95%
Cadmium, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 1m coil, thickness 0.5mm, as rolled, 100%
Cadmium, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%
Cadmium, foil, 10mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, wire reel, 0.2m, diameter 1.0mm, hard, 99.999%
Cadmium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, 25mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 50x100mm, thickness 1.0mm, as rolled, 99.999%
Cadmium, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95%
Cadmium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%
Cadmium, foil, 15mm disks, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%
Cadmium, foil, 50x200mm, thickness 1.0mm, as rolled, 99.999%
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cadmium, foil, 15mm disks, thickness 0.025mm, 99.7+%
Cadmium, foil, 15mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, foil, 10mm disks, thickness 0.25mm, as rolled, 99.9%
Cadmium, wire reel, 0.2m, diameter 0.5mm, hard, 99.99+%