Merck
  • Oral collagen-derived dipeptides, prolyl-hydroxyproline and hydroxyprolyl-glycine, ameliorate skin barrier dysfunction and alter gene expression profiles in the skin.
Oral collagen-derived dipeptides, prolyl-hydroxyproline and hydroxyprolyl-glycine, ameliorate skin barrier dysfunction and alter gene expression profiles in the skin.
Biochemical and biophysical research communications (2014-12-17)
Jun Shimizu, Naoto Asami, Aya Kataoka, Fumihito Sugihara, Naoki Inoue, Yoshifumi Kimira, Masahiro Wada, Hiroshi Mano
ABSTRACT
Oral supplementation with collagen hydrolysate (CH) has been shown to improve the condition of the skin in humans and experimental animals. Several hydroxyproline-containing oligo-peptides were previously detected in human peripheral blood after the ingestion of CH, and the two dipeptides, prolyl-hydroxyproline (PO) and hydroxyprolyl-glycine (OG), have been proposed to have beneficial effects on human health. When HR-1 hairless mice were fed a HR-AD diet, which lacked magnesium and zinc, transepidermal water loss (TEWL) increased and water content of stratum corneum decreased. In the present study, we investigated the effects of dietary PO and OG on skin barrier dysfunction in HR-1 hairless mice. Mice were fed a HR-AD diet with or without PO (0.15%) and OG (0.15%) for 35 consecutive days. The administration of PO and OG significantly decreased TEWL, and significantly increased water content of stratum corneum. A DNA microarray analysis of the dorsal skin revealed differences in gene expression between the group administered PO and OG and the control group. We also identified muscle-related Gene Ontology as a result of analyzing the up-regulated genes. These results suggested that the administration of PO and OG improved skin barrier dysfunction and altered muscle-related gene expression.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture