Merck
  • Net clinical benefit of antithrombotic therapy in patients with atrial fibrillation and chronic kidney disease: a nationwide observational cohort study.
Net clinical benefit of antithrombotic therapy in patients with atrial fibrillation and chronic kidney disease: a nationwide observational cohort study.
Journal of the American College of Cardiology (2014-12-17)
Anders Nissen Bonde, Gregory Y H Lip, Anne-Lise Kamper, Peter Riis Hansen, Morten Lamberts, Kristine Hommel, Morten Lock Hansen, Gunnar Hilmar Gislason, Christian Torp-Pedersen, Jonas Bjerring Olesen
ABSTRACT
The balance between stroke reduction and increased bleeding associated with antithrombotic therapy among patients with atrial fibrillation (AF) and chronic kidney disease (CKD) is controversial. This study assessed the risk associated with CKD in individual CHA₂DS₂-VASc (Congestive heart failure; Hypertension; Age ≥75 years; Diabetes mellitus; previous Stroke, transient ischemic attack, or thromboembolism; Vascular disease; Age 65 to 74 years; Sex category) strata and the net clinical benefit of warfarin in patients with AF and CKD in a nationwide cohort. By individual-level linkage of nationwide Danish registries, we identified all patients discharged with nonvalvular AF from 1997 to 2011. The stroke risk associated with non-end-stage CKD and end-stage CKD (e.g., patients on renal replacement therapy [RRT]) was estimated using Cox regression analyses. The net clinical benefit of warfarin was assessed using 4 endpoints: a composite endpoint of death/hospitalization from stroke/bleeding; a composite endpoint of fatal stroke/fatal bleeding; cardiovascular death; and all-cause death. From nonvalvular AF patients (n = 154,259), we identified 11,128 patients (7.2%) with non-end-stage CKD and 1,728 (1.1%) receiving RRT. In all CHA₂DS₂-VASc risk groups, RRT was independently associated with a higher risk of stroke/thromboembolism, from a 5.5-fold higher risk in patients with CHA₂DS₂-VASc score = 0 to a 1.6-fold higher risk in patients with CHA₂DS₂-VASc score ≥2. In patients receiving RRT with CHA₂DS₂-VASc score ≥2, warfarin was associated with lower risk of all-cause death (hazard ratio [HR]: 0.85, 95% confidence interval [CI]: 0.72 to 0.99). In non-end-stage CKD patients with CHA₂DS₂-VASc score ≥2, warfarin was associated with a lower risk of a composite outcome of fatal stroke/fatal bleeding (HR: 0.71, 95% CI: 0.57 to 0.88), a lower risk of cardiovascular death (HR: 0.80, 95% CI: 0.74 to 0.88), and a lower risk of all-cause death (HR: 0.64, 95% CI: 0.60 to 0.69). CKD is associated with a higher risk of stroke/thromboembolism across stroke risk strata in AF patients. High-risk CKD patients (CHA₂DS₂-VASc ≥2) with AF benefit from warfarin treatment for stroke prevention.
Product Description
Supelco
Warfarin, PESTANAL®, analytical standard
Supelco
Warfarin, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Aspirin, meets USP testing specifications
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material