Merck
  • Adiponectin induces apoptosis in hepatocellular carcinoma through differential modulation of thioredoxin proteins.
Adiponectin induces apoptosis in hepatocellular carcinoma through differential modulation of thioredoxin proteins.
Biochemical pharmacology (2014-12-17)
Su-Qian Xing, Chen-Guang Zhang, Ji-Fang Yuan, Hui-Min Yang, Shu-Dong Zhao, Hong Zhang
ABSTRACT
Adiponectin blocks hepatocellular carcinoma (HCC) progression by inducing cell apoptosis through the modulation of C-Jun N-terminal kinase and mammalian target of rapamycin. However, the precise upstream signaling pathways or molecules remain elusive. In the present study, we analyzed the role of antioxidant protein thioredoxin (Trx) in adiponectin-induced apoptosis in HCC. Adiponectin treatment decreased the viabilities of both HepG2 and Huh7 HCC cells accompanied by increased accumulation of intracellular reactive oxygen species, as evidenced by 2',7'-dichlorodihydrofluorescein diacetate staining. Pretreatment of these cells with the deoxidant N-acetylcysteine blocked the inhibitory effect of adiponectin. Levels of Trx2 protein in both HCC cells were significantly decreased, and the level of Trx1 was significantly inhibited in Huh7 cells while unchanged in HepG2 cells. However, the redox state of Trx1 was altered from reduced to the oxidized form following adiponectin treatment in HepG2 cells. Overexpression of both Trx proteins rescued adiponectin-induced cell apoptosis, whereas mutated Trx proteins were less effective. Further analysis suggested that both ASK1 and JNK signaling are involved in this process. Trx1 and Trx2 proteins also manifested protective effects on HCC cells in response to adiponectin treatment in a xenograft tumor model. Furthermore, high levels of Trx proteins and low adiponectin expression levels were found in primary human HCC samples compared with paracancerous tissues. These results suggest that Trx proteins play important roles in mediating adiponectin-induced HCC cell apoptosis, thus providing new insights into the pathogenesis of HCC and identifying adiponectin and Trx proteins as potential combinational therapeutic targets for the treatment of HCC.
Product Description
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
Sigma-Aldrich
Okadaic acid from Prorocentrum concavum, 92-100% (HPLC)
Sigma-Aldrich
Iodoacetic acid, ≥98.0% (T)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Thioredoxin from Escherichia coli, recombinant, expressed in E. coli, essentially salt-free, lyophilized powder, ≥3 units/mg protein
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material