Merck
  • Effects of anodic titanium dioxide nanotubes of different diameters on macrophage secretion and expression of cytokines and chemokines.
Effects of anodic titanium dioxide nanotubes of different diameters on macrophage secretion and expression of cytokines and chemokines.
Cell proliferation (2014-12-19)
W L Lü, N Wang, P Gao, C Y Li, H S Zhao, Z T Zhang
ABSTRACT
To investigate effects of TiO2 nanotubes of different diameters on J744A.1 macrophage behaviour, secretion and expression of pro-inflammatory cytokines and chemokines. Macrophage-like J744A.1 cells were cultured on three types of Ti surface: mechanically polished titanium plus 30 and 80 nm TiO2 nanotube surfaces, for 4, 24 and 48 h. Macrophage adhesion and proliferation were assessed using CCK-8 assay. Levels of pro-inflammatory cytokines (TNF-α, IL-1β and IL-6) and chemokines (MCP-1 and MIP-1α) secreted into the supernatant were measured using the Cytometric Bead Arrays kit. TNF-α, MCP-1 and MIP-1α gene expression were quantitatively analysed by real-time PCR. These show that TiO2 nanotube surfaces, especially of 80 nm TiO2 nanotube, benefited macrophage adhesion and proliferation, and reduced protein secretion and mRNA expression of TNF-α, MCP-1 and MIP-1α. IL-1β and IL-6 were undetectable on all the surfaces at all times. TiO2 nanotube surfaces, especially of 80 nm TiO2 nanotube, reduced inflammatory response in vitro, which might be part of a basis for rapid osseointegration in implants with TiO2 nanotube surfaces in animal studies.
Product Description
Titanium, wire reel, 200m, diameter 0.20mm, annealed, 99.6+%
Titanium, wire reel, 5m, diameter 0.25mm, as drawn, 99.99+%
Titanium, wire reel, 2m, diameter 1.0mm, as drawn, 99.99+%
Titanium, wire reel, 200m, diameter 0.125mm, annealed, 99.6+%
Titanium, wire, straight, 1000mm, diameter 1.0mm, as drawn, 99.6+%
Titanium, wire reel, 500m, diameter 0.20mm, annealed, 99.6+%
Titanium, wire reel, 25m, diameter 0.25mm, annealed, 99.6+%
Titanium, wire reel, 5m, diameter 0.20mm, annealed, 99.6+%
Titanium, wire reel, 5m, diameter 1.0mm, as drawn, 99.6+%
Titanium, wire reel, 1m, diameter 0.5mm, as drawn, 99.99+%
Titanium, tube, 500mm, outside diameter 2.03mm, inside diameter 1.63mm, wall thickness 0.2mm, annealed, 99.6+%
Titanium, wire reel, 100m, diameter 0.75mm, as drawn, 99.6+%
Titanium, wire reel, 5m, diameter 0.5mm, as drawn, 99.6+%
Titanium, wire reel, 0.5m, diameter 1.0mm, annealed, 99.6+%
Titanium, tube, 500mm, outside diameter 0.51mm, inside diameter 0.35mm, wall thickness 0.08mm, annealed, 99.6+%
Titanium, wire reel, 1m, diameter 0.25mm, as drawn, 99.99+%
Titanium, tube, 500mm, outside diameter 0.81mm, inside diameter 0.61mm, wall thickness 0.1mm, as drawn, 99.6+%
Titanium, wire reel, 50m, diameter 0.75mm, as drawn, 99.6+%
Titanium, wire reel, 5m, diameter 0.75mm, as drawn, 99.6+%
Titanium, wire reel, 25m, diameter 1.0mm, as drawn, 99.6+%
Titanium, wire reel, 20m, diameter 0.125mm, annealed, 99.6+%
Titanium, tube, 500mm, outside diameter 3.0mm, inside diameter 1.5mm, wall thickness 0.75mm, annealed, 99.6+%
Titanium, wire reel, 200m, diameter 0.25mm, annealed, 99.6+%
Titanium, wire reel, 25m, diameter 0.25mm, as drawn, 99.6+%
Titanium, wire reel, 20m, diameter 0.05mm, hard, 99.8%
Titanium, wire reel, 25m, diameter 1.0mm, annealed, 99.6+%
Titanium, wire reel, 200m, diameter 0.25mm, as drawn, 99.6+%
Titanium, wire reel, 25m, diameter 0.20mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 25mm, annealed, 99.6+%
Titanium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.6+%