Merck
  • Organization of vagal afferents in pylorus: mechanoreceptors arrayed for high sensitivity and fine spatial resolution?
Organization of vagal afferents in pylorus: mechanoreceptors arrayed for high sensitivity and fine spatial resolution?
Autonomic neuroscience : basic & clinical (2014-03-25)
Terry L Powley, Cherie N Hudson, Jennifer L McAdams, Elizabeth A Baronowsky, Felecia N Martin, Jacqueline K Mason, Robert J Phillips
ABSTRACT
The pylorus is innervated by vagal mechanoreceptors that project to gastrointestinal smooth muscle, but the distributions and specializations of vagal endings in the sphincter have not been fully characterized. To evaluate their organization, the neural tracer dextran biotin was injected into the nodose ganglia of rats. Following tracer transport, animals were perfused, and their pylori and antra were prepared as whole mounts. Specimens were processed to permanently label the tracer, and subsets were counterstained with Cuprolinic blue or immunostained for c-Kit. Intramuscular arrays (IMAs) in the circular muscle comprised the principal vagal afferent innervation of the sphincter. These pyloric ring IMAs were densely distributed and evidenced a variety of structural specializations. Morphometric comparisons between the arbors innervating the pylorus and a corresponding sample of IMAs in the adjacent antral circular muscle highlighted that sphincter IMAs branched profusely, forming more than twice as many branches as did antral IMAs (means of 405 vs. 165, respectively), and condensed their numerous neurites into compact receptive fields (∼48% of the area of antral IMAs) deep in the circular muscle (∼6μm above the submucosa). Separate arbors of IMAs in the sphincter interdigitated and overlapped to form a 360° band of mechanoreceptors encircling the pyloric canal. The annulus of vagal IMA arbors, putative stretch receptors tightly intercalated in the sphincter ring and situated near the lumen of the pyloric canal, creates an architecture with the potential to generate gut reflexes on the basis of pyloric sensory maps of high sensitivity and fine spatial resolution.
Sigma-Aldrich
D.P.X., Neutral mounting medium
Pricing and availability is not currently available.