Merck
  • Targeted inhibition of cell-surface serine protease Hepsin blocks prostate cancer bone metastasis.
Targeted inhibition of cell-surface serine protease Hepsin blocks prostate cancer bone metastasis.
Oncotarget (2014-03-25)
Xi Tang, Sumit S Mahajan, Liem T Nguyen, François Béliveau, Richard Leduc, Julian A Simon, Valeri Vasioukhin
ABSTRACT
The development of effective therapies inhibiting prostate cancer progression and metastasis may substantially impact prostate cancer mortality and potentially reduce the rates of invasive treatments by enhancing the safety of active surveillance strategies. Hepsin (HPN) is a cell surface serine protease amplified in a subset of human sarcomas (7.2%), as well as in ovarian (10%), lung adeno (5.4%), lung squamous cell (4.5%), adenoid cystic (5%), breast (2.6%), uterine (1.7%) and colon (1.4%) carcinomas. While HPN is not amplified in prostate cancer, it is one of the most prominently overexpressed genes in the majority of human prostate tumors and genetic experiments in mice indicate that Hepsin promotes prostate cancer metastasis, particularly metastasis to the bone marrow. We report here the development, analysis and animal trial of the small-molecule Hepsin inhibitor HepIn-13. Long-term exposure to HepIn-13 inhibited bone, liver and lung metastasis in a murine model of metastatic prostate cancer. These findings indicate that inhibition of Hepsin with small-molecule compounds could provide an effective tool for attenuation of prostate cancer progression and metastasis.
Product Description
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Serine, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%