Merck
  • Proteomics-based identification of the tumor suppressor role of aminoacylase 1 in hepatocellular carcinoma.
Proteomics-based identification of the tumor suppressor role of aminoacylase 1 in hepatocellular carcinoma.
Cancer letters (2014-05-23)
Xuyong Wei, Jie Li, Haiyang Xie, Qi Ling, Jianguo Wang, Di Lu, Lin Zhou, Xiao Xu, Shusen Zheng
ABSTRACT
The present work aimed to investigate the expression and role of aminoacylase 1 (ACY1) in hepatocellular carcinoma (HCC) based on a proteomic study. The study results revealed that the expression of ACY1 was much lower in HCC tissues. ACY1 expression significantly correlated with the serum alpha fetoprotein level and tumor invasiveness. The knockdown of ACY1 in SMMC7721 cells promoted cell viability and invasiveness. In contrast, the restoration of ACY1 in BEL7402 cells inhibited cell viability and invasiveness. Further studies indicated that the knockdown of ACY1 increased the protein expression of transforming growth factor beta 1 and extracellular signal-regulated kinase 1 expression. The study's results indicated that ACY1 acts as a tumor suppressor in HCC.
Sigma-Aldrich
MISSION® esiRNA, targeting human CHN1
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Supelco
L-Lysine, analytical standard
Sigma-Aldrich
MISSION® esiRNA, targeting human ACY1, ABHD14A-ACY1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Chn1
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Acy1