Merck
  • Sustained delivery of latanoprost by thermosensitive chitosan-gelatin-based hydrogel for controlling ocular hypertension.
Sustained delivery of latanoprost by thermosensitive chitosan-gelatin-based hydrogel for controlling ocular hypertension.
Acta biomaterialia (2014-06-11)
Yung-Hsin Cheng, Kuo-Hsuan Hung, Tung-Hu Tsai, Chia-Jung Lee, Ruy-Yu Ku, Allen Wen-Hsiang Chiu, Shih-Hwa Chiou, Catherine Jui-Ling Liu
ABSTRACT
Glaucoma is an irreversible ocular disease that may lead to progressive visual field loss and eventually to blindness with inadequately controlled intraocular pressure (IOP). Latanoprost is one of the most potent ocular hypotensive compounds, the current first-line therapy in glaucoma. However, the daily instillation required for efficacy and undesirable side-effects are major causes of treatment adherence failure and persistence in glaucoma therapy. In the present study, we developed an injectable thermosensitive chitosan/gelatin/glycerol phosphate (C/G/GP) hydrogel as a sustained-release system of latanoprost for glaucoma treatment. The latanoprost-loaded C/G/GP hydrogel can gel within 1min at 37°C. The results show a sustained release of latanoprost from C/G/GP hydrogel in vitro and in vivo. The latanoprost-loaded C/G/GP hydrogel showed a good in vitro and in vivo biocompatibility. A rabbit model of glaucoma was established by intravitreal injection of triamcinolone acetonide. After a single subconjunctival injection of latanoprost-loaded C/G/GP hydrogel, IOP was significantly decreased within 8days and then remained at a normal level. The results of the study suggest that latanoprost-loaded C/G/GP hydrogel may have a potential application in glaucoma therapy.
Sigma-Aldrich
Latanoprost, ≥98% (HPLC)
Sigma-Aldrich
Formalin solution, neutral buffered, 10%, histological tissue fixative
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and 15 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
β-Glycerophosphate disodium salt hydrate, ≤1.0 mol % L-α-isomer