Merck
  • Bone regeneration in a massive rat femur defect through endochondral ossification achieved with chondrogenically differentiated MSCs in a degradable scaffold.
Bone regeneration in a massive rat femur defect through endochondral ossification achieved with chondrogenically differentiated MSCs in a degradable scaffold.
Biomaterials (2014-06-24)
Noriko Harada, Yoshinobu Watanabe, Kenji Sato, Satoshi Abe, Katsuyuki Yamanaka, Yuhiro Sakai, Tadashi Kaneko, Takashi Matsushita
ABSTRACT
Mesenchymal stem cells (MSCs) are multipotent cells capable of proliferating and differentiating into several lineages. In regenerative medicine, their potential as a resource for tissue-replacement therapy is receiving much attention. However, transplanting MSCs to repair larger bone defects in animal models has so far proved disappointing. Here we report on the healing of both critical-sized (5 mm) and massive (15 mm) full-thickness femur defects in rats by implanting a uniquely fabricated PLGA scaffold seeded with MSCs pre-differentiated in vitro into cartilage-forming chondrocytes (MSC-DCs). This strategy closely mimics endochondral ossification, the process by which long bones develop in nature. It is thought that because the transplanted MSC-DCs induced natural bone formation, the defect size was not critical to the outcome. Crucially, after 8 weeks the mean biomechanical strength of femora with the massive 15 mm implant reached 75% that of a normal rat femur, while in the case of 5 mm implants there was no significant difference. Successful healing was also highly reproducible, with bone union occurring in all treated animals examined radiologically 8 or 16 weeks after surgery.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
1,4-Dioxane, analytical standard
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material