Merck
  • Systemic gene dysregulation in classical Galactosaemia: Is there a central mechanism?
Systemic gene dysregulation in classical Galactosaemia: Is there a central mechanism?
Molecular genetics and metabolism (2014-09-02)
K P Coss, E P Treacy, E J Cotter, I Knerr, D W Murray, Y S Shin, P P Doran
ABSTRACT
Classical Galactosaemia is a rare disorder of carbohydrate metabolism caused by a deficiency of galactose-1-phosphate uridyltransferase (GALT). The disease is life-threatening in the neonate, and the only treatment option is life-long dietary restriction of galactose. However, long-term complications persist in treated patients including cognitive impairments, speech and language abnormalities and premature ovarian insufficiency in females. Microarray analysis of T-lymphocytes from treated adult patients identified systemic dysregulation of numerous gene pathways, including the glycosylation, inflammatory and inositol pathways. Analysis of gene expression in patient-derived dermal fibroblasts of patients exposed to toxic levels of galactose, with immunostaining, has further identified the susceptibility of the glycosylation gene alpha-1,2-mannosyltransferase (ALG9) and the inflammatory gene annexin A1 (ANXA1) to increased galactose concentrations. These data suggest that Galactosaemia is a multi-system disorder affecting numerous signalling pathways.
Product Description
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Nutrient Mixture F-10 Ham, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
USP
Galactose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Galactose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)