Merck
  • Synthesis and liquid crystalline properties of new diols containing azomethine groups.
Synthesis and liquid crystalline properties of new diols containing azomethine groups.
Molecules (Basel, Switzerland) (2010-07-27)
Issam Ahmed Mohammed, Govindarajan Sankar, Melati Khairuddean, Abu Bakar Mohamad
ABSTRACT
A series of new mesogenic azomethine diols were successfully synthesized by condensation reactions between various chloroalkanols and N,N'-bis(4-hydroxy)-benzylidene-o-toluidine (1). The structures of these compounds were confirmed by CHN, FT-IR, (1)H-NMR, and (13)C-NMR spectrophotometer. Their thermotropic liquid crystalline behavior was studied using differential scanning calorimetry (DSC) and polarizing optical microscope (POM). 4,4'-di(4-Hydroxybutoxy)-N-benzylidine-o-tolidine (2a) does not exhibit liquid crystalline properties. A nematic texture was observed for mesogenic diols 2b, and 2d, whereas the diol 2c exhibits a smectic mesophase. The increase of terminal alkyl chain in these mesogenic diols leads to a decrease in the transition temperature.
Sigma-Aldrich
8-Chloro-1-octanol, 98%
Pricing and availability is not currently available.