Merck
  • Ytterbium Trifluoromethanesulfonate Mediated Cross-Aldol Reaction between Ketones and Aldehydes.
Ytterbium Trifluoromethanesulfonate Mediated Cross-Aldol Reaction between Ketones and Aldehydes.
Bulletin of the Chemical Society of Japan (1994)
Fukuzawa S-I, et al.