Merck
Superoxide-mediated decomposition of biological S-nitrosothiols.
The Journal of biological chemistry (1998-04-16)
S Aleryani, E Milo, Y Rose, P Kostka
ABSTRACT
Incubation of S-nitrosocysteine or S-nitrosoglutathione (5-100 M) in the presence of a generator of superoxide (xanthine/xanthine oxidase) resulted in a time-dependent decomposition of S-nitrosothiols and accumulation of nitrite/nitrate in reaction mixtures. Quantitatively, the amounts of nitrite/nitrate represented >90% of nitrosonium equivalent of S-nitrosothiols degraded during the incubation. The reaction rates were unaffected by the presence catalase (1 unit/ml). Kinetic analysis showed that the degradation of S-nitrosothiols in the presence of superoxide proceeded at second order rate constants of 76,900 M-1 s-1 (S-nitrosocysteine) and 12,800 M-1 s-1 (S-nitrosoglutathione), respectively, with a stoichiometric ratio of 1 mol of S-nitrosothiol per 2 mol of superoxide. The findings provide the evidence for the involvement of superoxide in the metabolism of S-nitrosothiols. Furthermore, substantially slower reaction rates of superoxide with S-nitrosothiols relative to the reaction rate with NO are consistent with the contention that the transient formation of S-nitrosothiols in biological systems may protect NO from its rapid destruction by superoxide, thus enabling these compounds to serve as carriers or buffers of NO.
Sigma-Aldrich
Ammonium sulfamate, ACS reagent, ≥98.0%
Pricing and availability is not currently available.