Merck
  • The properties of molecular fragments obtained on treating calfskin collagen with collagenase from Clostridium histolyticum.
The properties of molecular fragments obtained on treating calfskin collagen with collagenase from Clostridium histolyticum.
European journal of biochemistry (1968-12-05)
M Stark, K Kühn
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder (from 0.2 μm filtered solution), suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
Roche
Collagenase/Dispase®, lyophilized, non-sterile, optimum pH 7.0-8.0
Roche
Collagenase A, from Clostridium histolyticum