Merck
  • TRPM1 (Melastatin-1/MLSN1) mRNA expression in Spitz nevi and nodular melanomas.
TRPM1 (Melastatin-1/MLSN1) mRNA expression in Spitz nevi and nodular melanomas.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc (2009-04-28)
Lori A Erickson, Gary A Letts, Sonali M Shah, Jeffrey B Shackelton, Lyn M Duncan
ABSTRACT
The transient receptor potential cation channel, subfamily M, member 1 (TRPM1/Melastatin-1/MLSN-1) expression has been shown to have prognostic utility in the evaluation of primary cutaneous melanoma. We analyzed a series of spindled and epithelioid cell nevi (Spitz) and primary cutaneous nodular melanomas to determine whether the expression of TRPM1 mRNA may be useful in distinguishing between Spitz nevi and nodular melanomas and to further examine the patterns of TRPM1 mRNA expression in cutaneous melanocytic proliferations. Formalin-fixed, paraffin-embedded tissues from 95 Spitz nevi and 33 nodular melanomas were analyzed for the expression of TRPM1 mRNA by in situ hybridization using (35)S-labeled riboprobes. Ubiquitous melanocytic expression of TRPM1 mRNA was observed in 56 of 95 (59%) Spitz nevi and 4 of 33 (12%) nodular melanomas. Diffusely scattered loss of TRPM1 mRNA was identified in 38 of 95 (40%) Spitz nevi and 2 of 33 (6%) nodular melanomas. Regional loss of the TRPM1 mRNA expression by a significant subset of dermal tumor cells or a complete absence of TRPM1 expression by the dermal tumor was identified in 27 of 33 (82%) nodular melanomas, but only 1 of 95 (1%) Spitz nevi. These findings suggest that the pattern of TRPM1 mRNA expression may be helpful in the differentiation of Spitz nevi and nodular melanomas. Of the 16 patients who experienced metastasis, 15 (94%) had primary tumors that displayed reduced MLSN mRNA expression by all or a part of the dermal tumor.