Merck
  • Nucleation Kinetics, Growth and Characterization Studies of a Diamagnetic Crystal-Zinc Sulphate Heptahydrate (ZSHH).
Nucleation Kinetics, Growth and Characterization Studies of a Diamagnetic Crystal-Zinc Sulphate Heptahydrate (ZSHH).
Journal of Chemistry (2009)
Kanagadurai R, et al.