Merck
  • Development of classical Hodgkin's lymphoma in an adult with biallelic STXBP2 mutations.
Development of classical Hodgkin's lymphoma in an adult with biallelic STXBP2 mutations.
Haematologica (2012-10-27)
Maciej Machaczka, Monika Klimkowska, Samuel C C Chiang, Marie Meeths, Martha-Lena Müller, Britt Gustafsson, Jan-Inge Henter, Yenan T Bryceson
ABSTRACT
Experimental model systems have delineated an important role for cytotoxic lymphocytes in the immunosurveillance of cancer. In humans, perforin-deficiency has been associated with occurrence of hematologic malignancies. Here, we describe an Epstein-Barr virus-positive classical Hodgkin's lymphoma in a patient harboring biallelic mutations in STXBP2, a gene required for exocytosis of perforin-containing lytic granules and associated with familial hemophagocytic lymphohistocytosis. Cytotoxic T lymphocytes were found infiltrating the tumor, and a high frequency of Epstein-Barr virus-specific cytotoxic T lymphocytes were detected in peripheral blood. However, lytic granule exocytosis and cytotoxicity by cytotoxic T lymphocytes, as well as natural killer cells, were severely impaired in the patient. Thus, the data suggest a link between defective lymphocyte exocytosis and development of lymphoma in STXBP2-deficient patients. Therefore, with regards to treatment of familial hemophagocytic lymphohistocytosis patients with mutations in genes required for lymphocyte exocytosis, it is important to consider both the risks of hemophagocytic lymphohistocytosis and malignancy.