Merck
  • Electrophysiological and behavioral responses of Thanatophilus sinuatus Fabricius (Coleoptera: Silphidae) to selected cadaveric volatile organic compounds.
Electrophysiological and behavioral responses of <I>Thanatophilus sinuatus</I> Fabricius (Coleoptera: Silphidae) to selected cadaveric volatile organic compounds.
Journal of Forensic Sciences (2013)
Dekeirsschieter J, et al.