Merck
  • Electrochemical photocatalytic degradation of dye solution with a TiO2-coated stainless steel electrode prepared by electrophoretic deposition.
Electrochemical photocatalytic degradation of dye solution with a TiO<SUB>2</SUB>-coated stainless steel electrode prepared by electrophoretic deposition.
Applied Catalysis. B, Environmental (2013)
Lin WC, et al.