Merck
  • Lignans and neolignans from the stems of Vibrunum erosum and their neuroprotective and anti-inflammatory activity.
Lignans and neolignans from the stems of Vibrunum erosum and their neuroprotective and anti-inflammatory activity.
Archives of pharmacal research (2014-03-29)
Seo-Ji In, Kyeong-Hwa Seo, Na-Young Song, Dong-Sung Lee, Youn-Chul Kim, Nam-In Baek
ABSTRACT
A new lignan, (7'S,8S,8'S)-3,5'-dimethoxy-3',4,9'-trihydroxy-7',9-epoxy-8,8'-lignan, named vibruresinol (1), was isolated from the stems of Viburnum erosum by silica gel, ODS, and Sephadex LH-20 column chromatography. In addition, four other known lignans, (7'R,8S,8'S)-3,5'-dimethoxy-3',4,8',9'-tetrahydroxy-7',9-epoxy-8,8'-lignan (2), (+)-syringaresinol (3), (+)-pinoresinol (4), and (+)-pinoresinol-4-O-β-D-glucopyranoside (5), and five known neolignans, herpetol (6), vibsanol (7), (-)-dehydrodiconiferyl alcohol (8), icariside E4 (9), and dihydrodehydrodiconiferyl alcohol (10), were isolated in the same manner. The chemical structures of the compounds were determined based on spectroscopic data including NMR, MS, and IR. All of the compounds described above were isolated from V. erosum for the first time. The isolated compounds 3, 4, and 6 were evaluated for neuroprotective activity on glutamate-induced cell death in HT22 cells and had EC50 values of 6.33 ± 1.22, 6.96 ± 0.65, and 9.15 ± 0.36 μM, respectively. Likewise, the same compounds had inhibitory activity on NO production in LPS-induced RAW 264.7 cells with IC50 values of 8.30 ± 1.56, 7.89 ± 1.22, and 9.32 ± 0.36 μM, respectively.
Product Description
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC