Merck
  • Preparation and evaluation of submicron-carriers for naringenin topical application.
Preparation and evaluation of submicron-carriers for naringenin topical application.
International journal of pharmaceutics (2015-01-24)
Ming-Jun Tsai, Yaw-Bin Huang, Jhih-Wun Fang, Yaw-Syan Fu, Pao-Chu Wu
ABSTRACT
Submicron emulsion system is one kind of submicron-carrier that can ensure close contact and increase the amount of drug transport into the skin. In the present study, naringenin was loaded into a submicron emulsion system for topical applications. The enhancement effect of drug permeability through skin, stability, and skin irritation of naringenin-loaded submicron emulsions were evaluated. The results showed that the transdermal amount and deposition amount in skin of naringenin from submicron emulsion formulations were significantly increased when compared to the control group of saturated aqueous solution of naringenin. The drug-loaded submicron emulsions showed thermodynamic stability after centrifugation and cooling-heating cycle tests. The level of drug was more than 98% after 3 months of storage at 25°C and 40°C. In skin irritation test, the result also demonstrated that naringenin-loaded submicron emulsion had less skin irritation, indicating that the formulation can possibly be developed for topical application.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hesperetin, ≥95%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Hesperetin, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Naringenin, natural (US), 98%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Isopropyl myristate, ≥98%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Isopropyl myristate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Isopropyl myristate, 98%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
(±)-Naringenin, ≥95%