Merck
  • Potent inhibition of macrophage migration inhibitory factor (MIF) by myeloperoxidase-dependent oxidation of epicatechins.
Potent inhibition of macrophage migration inhibitory factor (MIF) by myeloperoxidase-dependent oxidation of epicatechins.
The Biochemical journal (2014-06-12)
Nina Dickerhof, Nicholas J Magon, Joel D A Tyndall, Anthony J Kettle, Mark B Hampton
ABSTRACT
MIF (macrophage migration inhibitory factor) plays a central role in the promotion and maintenance of the inflammatory response. It is implicated in a number of inflammatory diseases including sepsis, arthritis and colitis, and in diseases with an inflammatory component, such as atherosclerosis, diabetes and cancer. MIF has an unusual N-terminal proline with catalytic activity, and targeting of this residue by small-molecule inhibitors has been shown to interfere with the biological activity of MIF. The objective of the present study was to determine if MIF was susceptible to modification by epicatechins, a group of dietary flavonoids with known anti-inflammatory properties. Epicatechins are substrates for peroxidases including neutrophil-derived MPO (myeloperoxidase). In the present study we show that oxidation of the catechol moiety of epicatechins to an ο-quinone by MPO generates potent MIF inhibitors. Near complete inhibition of MIF by the MPO/H2O2/epicatechin system was achieved at equimolar concentrations of epicatechin and MIF, even in the presence of other MPO substrates. We have characterized the modification introduced by oxidized (-)-epicatechin on MIF by LC-MS (liquid chromatography MS) and found it to occur at the N-terminal proline. We propose that MIF inhibition by oxidized epicatechins contributes to the anti-inflammatory activity of these compounds.
Sigma-Aldrich
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Sodium (meta)periodate, ≥99.0%
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Uric acid, ≥99%, crystalline
SAFC
L-Methionine
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Epigallocatechin gallate, primary reference standard
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(−)-Epigallocatechin gallate, analytical standard
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
Sodium periodate, ACS reagent, ≥99.8%
Supelco
L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Iodoacetamide
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland