Merck
  • Basal cells contribute to innate immunity of the airway epithelium through production of the antimicrobial protein RNase 7.
Basal cells contribute to innate immunity of the airway epithelium through production of the antimicrobial protein RNase 7.
Journal of immunology (Baltimore, Md. : 1950) (2015-02-26)
Gimano D Amatngalim, Yolanda van Wijck, Yvonne de Mooij-Eijk, Renate M Verhoosel, Jürgen Harder, Annemarie N Lekkerkerker, Richard A J Janssen, Pieter S Hiemstra
ABSTRACT
Basal cells play a critical role in the response of the airway epithelium to injury and are recently recognized to also contribute to epithelial immunity. Antimicrobial proteins and peptides are essential effector molecules in this airway epithelial innate immunity. However, little is known about the specific role of basal cells in antimicrobial protein and peptide production and about the regulation of the ubiquitous antimicrobial protein RNase 7. In this study, we report that basal cells are the principal cell type producing RNase 7 in cultured primary bronchial epithelial cells (PBEC). Exposure of submerged cultured PBEC (primarily consisting of basal cells) to the respiratory pathogen nontypeable Haemophilus influenzae resulted in a marked increase in expression of RNase 7, although this was not observed in differentiated air-liquid interface cultured PBEC. However, transient epithelial injury in air-liquid interface-cultured PBEC induced by cigarette smoke exposure led to epidermal growth factor receptor-mediated expression of RNase 7 in remaining basal cells. The selective induction of RNase 7 in basal cells by cigarette smoke was demonstrated using confocal microscopy and by examining isolated luminal and basal cell fractions. Taken together, these findings demonstrate a phenotype-specific innate immune activity of airway epithelial basal cells, which serves as a second line of airway epithelial defense that is induced by airway epithelial injury.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Sodium chloride solution, 5 M
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O