Merck
  • Glucose and insulin administration while maintaining normoglycemia inhibits whole body protein breakdown and synthesis after cardiac surgery.
Glucose and insulin administration while maintaining normoglycemia inhibits whole body protein breakdown and synthesis after cardiac surgery.
Journal of applied physiology (Bethesda, Md. : 1985) (2014-09-27)
Roupen Hatzakorzian, Dominique Shum-Tim, Linda Wykes, Ansgar Hülshoff, Helen Bui, Evan Nitschmann, Ralph Lattermann, Thomas Schricker
ABSTRACT
We investigated the effect of insulin administered as part of a hyperinsulinemic-normoglycemic clamp on protein metabolism after coronary artery bypass grafting (CABG) surgery. Eighteen patients were studied, with nine patients in the control group receiving standard metabolic care and nine patients receiving insulin (5 mU·kg(-1)·min(-1)). Whole body glucose production, protein breakdown, synthesis, and oxidation were determined using stable isotope tracer kinetics (l-[1-(13)C]leucine, [6,6-(2)H2]glucose) before and 6 h after the procedure. Plasma amino acids, cortisol, and lactate were also measured. Endogenous glucose production (preoperatively 10.0 ± 1.6, postoperatively 3.7 ± 2.5 μmol·kg(-1)·min(-1); P = 0.0001), protein breakdown (preoperatively 105.3 ± 9.8, postoperatively 85.2 ± 9.2 mmol·kg(-1)·h(-1); P = 0.0005) and synthesis (preoperatively 88.7 ± 8.7, postoperatively 72.4 ± 8.4 mmol·kg(-1)·h(-1); P = 0.0005) decreased in the presence of hyperinsulinemia, whereas both parameters remained unchanged in the control group. A positive correlation between endogenous glucose production and protein breakdown was observed in the insulin group (r(2) = 0.385). Whole body protein oxidation and balance decreased after surgery in patients receiving insulin without reaching statistical significance. In the insulin group the plasma concentrations of 13 of 20 essential and nonessential amino acids decreased to a significantly greater extent than in the control group. In summary, supraphysiological hyperinsulinemia, while maintaining normoglycemia, decreased whole body protein breakdown and synthesis in patients undergoing CABG surgery. However, net protein balance remained negative.
Product Description
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Leucine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
DL-Leucine, ≥99% (HPLC)
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard