Merck
  • Osmoprotectants suppress the production and activity of matrix metalloproteinases induced by hyperosmolarity in primary human corneal epithelial cells.
Osmoprotectants suppress the production and activity of matrix metalloproteinases induced by hyperosmolarity in primary human corneal epithelial cells.
Molecular vision (2014-10-30)
Ruzhi Deng, Zhitao Su, Xia Hua, Zongduan Zhang, De-Quan Li, Stephen C Pflugfelder
ABSTRACT
Hyperosmolarity has been recognized as a proinflammatory stress in the pathogenesis of dry eye disease. This study investigated the suppressive effect of osmoprotectants (L-carnitine, erythritol, and betaine) on the production and activity of matrix metalloproteinases (MMPs) in primary human corneal epithelial cells (HCECs) exposed to hyperosmotic stress. Primary HCECs were established from fresh donor limbal tissue explants. The cultures in iso-osmolar medium (312 mOsM) were switched to hyperosmotic media with or without prior incubation with different concentrations of L-carnitine, erythritol, or betaine (2, 10, or 20 mM). The mRNA expression of the MMPs was determined with reverse transcription and quantitative real-time PCR (RT-qPCR). Protein production and activity were evaluated with immunofluorescent staining and gelatin zymography. Hyperosmotic media (400, 450, or 500 mOsM) significantly stimulated mRNA expression of collagenase MMP-13, gelatinases MMP-9 and MMP-2, stromelysin MMP-3, and matrilysin MMP-7, mostly in an osmolarity-dependent fashion. The stimulated mRNA expression and protein production of these MMPs were significantly but differentially suppressed by L-carnitine, erythritol, or betaine, as evaluated with RT-qPCR and immunofluorescent staining. Interestingly, these osmoprotectants not only suppressed production but also inhibited activation of MMP-9 and MMP-2, as evaluated with gelatin zymography. Our findings for the first time demonstrate that osmoprotectants, L-carnitine, erythritol, and betaine, suppress the gene expression, protein production, and enzymatic activity of MMPs in HCECs exposed to hyperosmotic stress. L-carnitine appears to have the broadest and strongest suppressive effect on these MMPs. These osmoprotectants may have potential effects in protecting ocular surface epithelia from MMP-mediated disorders in dry eye disease.
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Levocarnitine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
meso-Erythritol, ≥99% (GC)
USP
Levocarnitine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Carnitine inner salt, synthetic, ≥98%
Erythritol, European Pharmacopoeia (EP) Reference Standard
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent