Merck
  • A new optimized formulation of cationic solid lipid nanoparticles intended for gene delivery: development, characterization and DNA binding efficiency of TCERG1 expression plasmid.
A new optimized formulation of cationic solid lipid nanoparticles intended for gene delivery: development, characterization and DNA binding efficiency of TCERG1 expression plasmid.
International journal of pharmaceutics (2014-07-08)
Anna Fàbregas, Noemí Sánchez-Hernández, Josep Ramon Ticó, Encarna García-Montoya, Pilar Pérez-Lozano, Josep M Suñé-Negre, Cristina Hernández-Munain, Carlos Suñé, Montserrat Miñarro
ABSTRACT
Solid lipid nanoparticles (SLNs) are being considered as a new approach for therapeutics for many known diseases. In addition to drug delivery, their use as non-viral vectors for gene delivery can be achieved by the inclusion of cationic lipids, which provide a positive surface potential that favours binding to the DNA backbone. This work is based on the idea that the optimization of the components is required as the first step in simplifying the qualitative and quantitative composition of SLNs as much as possible without affecting the essential properties that define SLNs as optimal non-viral vectors for gene delivery. We selected the best lipids and surfactants in terms of particle size and zeta potential and characterized the properties of the resulting nanoparticles using X-ray photoelectron spectroscopy (XPS) and atomic force microscopy (AFM). The SLNs had a particle size of approximately 120 nm and a positive surface charge of 42 mV. In addition, we analysed the main physicochemical characteristics of the bulk components of the nanoparticles using X-ray diffraction (XRD), differential scanning calorimetry (DSC) and mass spectrometry (MS). The suitability of the optimized SLNs for DNA binding was evaluated after the lyophilisation process using a carboxyl-terminal region of the TCERG1 gene, a human factor that has been implicated in several diseases. We show that the SLNs presented high efficiency in the binding of DNA, and importantly, they presented no toxicity when assayed in an in vivo system.
Sigma-Aldrich
Octadecylamine, ≥99% (GC)
Sigma-Aldrich
Octadecylamine, technical grade, 90%
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Octadecylamine, ≥99.0% (GC)
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Supelco
Stearic acid, analytical standard
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Stearic acid, United States Pharmacopeia (USP) Reference Standard