Merck
Extracorporeal photopheresis promotes IL-1β production.
Journal of immunology (Baltimore, Md. : 1950) (2015-02-15)
Erhan Yakut, Christopher Jakobs, Adriana Peric, Gabriela Michel, Nelli Baal, Gregor Bein, Bernhard Brüne, Veit Hornung, Holger Hackstein
ABSTRACT
Extracorporeal photopheresis (ECP) is a widely used clinical cell-based therapy exhibiting efficacy in heterogenous immune-mediated diseases such as cutaneous T cell lymphoma, graft-versus-host disease, and organ allograft rejection. Despite its documented efficacy in cancer immunotherapy, little is known regarding the induction of immunostimulatory mediators by ECP. In this article, we show that ECP promotes marked release of the prototypic immunostimulatory cytokine IL-1β. ECP primes IL-1β production and activates IL-1β maturation and release in the context of caspase-1 activation in monocytes and myeloid dendritic cells. Of interest, IL-1β maturation by ECP was fully intact in murine cells deficient in caspase-1, suggesting the predominance of an inflammasome-independent pathway for ECP-dependent IL-1β maturation. Clinically, patient analysis revealed significantly increased IL-1β production in stimulated leukapheresis concentrates and peripheral blood samples after ECP. Collectively, these results provide evidence for promotion of IL-1β production by ECP and offer new insight into the immunostimulatory capacity of ECP.
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Resiquimod, ≥98% (HPLC)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Supelco
Xanthotoxin, analytical standard
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Phenylacetic acid, 99%
Supelco
8-Methoxypsoralen, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)