Merck
  • Association between low education and higher global cardiovascular risk.
Association between low education and higher global cardiovascular risk.
Journal of clinical hypertension (Greenwich, Conn.) (2015-02-24)
Tiziana Di Chiara, Alessandra Scaglione, Salvatore Corrao, Christiano Argano, Antonio Pinto, Rosario Scaglione
ABSTRACT
This study was designed to evaluate the impact of educational status on global cardiovascular risk in a southern Italian urban population. The study population consisted of 488 consecutive outpatients aged 18 years and older. Educational status was categorized according to the number of years of formal education as follows: (1) low education group (<10 years) and (2) medium-high education group (10-15 years). In both groups, cardiometabolic comorbidities (obesity, visceral obesity, diabetes, dyslipidemia, metabolic syndrome, microalbuminuria, left ventricular hypertrophy) and global cardiovascular risk, according to international guidelines, were analyzed. Left ventricular mass index and ejection fraction by echocardiography and E/A ratio, by pulsed-wave Doppler, were calculated. The low education group was characterized by a significantly higher prevalence of patients with visceral obesity (P=.021), hypertension (P=.010), metabolic syndrome (P=.000), and microalbuminuria (P=.000) and greater global cardiovascular risk (P=.000). Significantly increased levels of microalbuminuria (P=.000) and significantly decreased values of E/A ratio (P=.000) were also detected in the low education group. Global cardiovascular risk correlated directly with waist-to-hip ratio (P=.010), microalbuminuria (P=.015), and the metabolic syndrome (P>.012) and inversely with educational status (P=.000). Education was independently (P=.000) associated with global cardiovascular risk. These data indicate a strong association between low education and cardiometabolic comorbidities suitable to influence the evolution of chronic degenerative diseases. Preventive strategies need to be more efficient and more effective in this patient population.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material