Merck
  • Development and validation of an on-line multidimensional SPE-LC-MS/MS method for the quantitation of Tetrandrine in blood samples.
Development and validation of an on-line multidimensional SPE-LC-MS/MS method for the quantitation of Tetrandrine in blood samples.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-03-10)
Sena Caglar, Rosa Morello, Karl-Siegfried Boos
ABSTRACT
On-line solid-phase extraction (SPE) is becoming an increasingly widespread technique in the clean-up of complex matrices such as body fluids, prior to chromatographic analysis. The use of small SPE columns instead of disposable SPE cartridges allows multiple injections and complete automation. In addition, it decreases the cost of consumables and improves the quality of the overall analysis. Coupling of SPE with HPLC combines sample preparation and separation in one system. In this paper a validated on-line multidimensional (MD) SPE-LC-MS/MS method is described for the determination of Tetrandrine (model drug) in human blood samples. The developed method showed the applicability of direct injection of plasma samples to an on-line MD-SPE-LC-MS/MS system to determine small molecules i.e. drugs. The experimental design is unique. Quantification was through tandem mass spectrometry with positive electrospray ionization (ESI) and multiple reactions monitoring (MRM). The limit of detection was calculated as 31.98 ng/mL. The linear range of the method was between 40.0 and 800.0 ng/mL. Pharmacokinetic parameters are usually determined by analysis of drug concentrations in plasma rather than whole blood. Parameters determined using plasma data may be misleading if concentrations of drug differ between plasma and red blood cells. We successfully applied the developed method for the determination of the distribution coefficient of the model drug Tetrandrine between human red blood cells and blood plasma proteins. The determination of distribution coefficient study results demonstrated that the developed method can provide direct and accurate measurement of RBC partitioning in a model drug and could be applied for screening of other compounds for potential high RBC partition, predicting potential drug toxicity and investigating mechanisms associated with RBC partitions.
Product Description
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%